Oxytetracycline hydrochloride, can be used as secondary standard'
Have Questions?
Catalog Number
Quantity
FSA301275000
500 mg
Catalog number FSA301275000
Price (MYR)
838.00
Add to cart
Quantity:
500 mg
Request bulk or custom format
Price (MYR)
838.00
Add to cart
Specifications
Chemical Name or MaterialOxytetracycline hydrochloride
TypeOxytetracycline Hydrochloride
Physical FormCrystalline Powder
CAS2058-46-0
Health Hazard 1LIGHT SENSITIVE