Acridine Orange, pure, ca. 55% dye content
Acridine Orange, pure, ca. 55% dye content
Acridine Orange, pure, ca. 55% dye content
Acridine Orange, pure, ca. 55% dye content
Thermo Scientific Chemicals
Acridine Orange, pure, ca. 55% dye content
CAS: 10127-02-3 | C17H20Cl3N3Zn | 438.10 g/mol
Have Questions?
Catalog NumberQuantity
FSA30091025025 g
Catalog number FSA300910250
Price (MYR)
626.00
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
626.00
Add to cart
Specifications
Chemical Name or MaterialAcridine Orange
Melting Point165°C
CAS10127-02-3
Health Hazard 1Warning
Health Hazard 2GHS H Statement
Harmful if swallowed. Harmful if inhaled. Harmful in contact with skin. Suspected of causing genetic defects.