Ethanol-d6, for NMR, anhydrous, 99% atom D, Thermo Scientific Chemicals
Ethanol-d6, for NMR, anhydrous, 99% atom D, Thermo Scientific Chemicals
Ethanol-d6, for NMR, anhydrous, 99% atom D, Thermo Scientific Chemicals
Ethanol-d6, for NMR, anhydrous, 99% atom D, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Ethanol-d6, for NMR, anhydrous, 99% atom D, Thermo Scientific Chemicals
CAS: 1516-08-1 | C2H6O | 52.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
1 mL
Catalog number FSA300680050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialEthanol-d6
Density0.8920g/mL
Deuterium (D)98.5% min.
Physical FormLiquid
Quantity5 mL
View more
For NMR
RUO – Research Use Only