5-Nitrosalicylic acid, 99%, Thermo Scientific Chemicals
5-Nitrosalicylic acid, 99%, Thermo Scientific Chemicals
5-Nitrosalicylic acid, 99%, Thermo Scientific Chemicals
5-Nitrosalicylic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
5-Nitrosalicylic acid, 99%, Thermo Scientific Chemicals
CAS: 96-97-9 | C7H5NO5 | 183.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass Bottle
Catalog number FSA300480050
Price (MYR)
282.00
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
282.00
Add to cart
Chemical Identifiers
CAS4039-32-1
IUPAC Namelithium(1+) bis(trimethylsilyl)azanide
Molecular FormulaC6H18LiNSi2
InChI KeyYNESATAKKCNGOF-UHFFFAOYSA-N
SMILES[Li+].C[Si](C)(C)[N-][Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Suppliers CofA≥98.0 to ≤102.0% (Titration)
FormCrystalline powder and/or chunks
Appearance (Color)White to yellow