Check Order Status
trans-2-Nonenal, 95%, Thermo Scientific™
Element Search
Structure Search
trans-2-Nonenal, 95%, Thermo Scientific™
Have Questions?
Catalog Number
Quantity
Packaging
FSA298310250
25 mL
Glass bottle
Catalog number
FSA298310250
Price (MYR)
746.00
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
746.00
Add to cart
Specifications
Chemical Name or Material
trans-2-Nonenal
Name Note
95%
CAS
18829-56-6
Health Hazard 1
GHS Signal Word:
Warning
Health Hazard 2
GHS H Statement