Diethyl acetylenedicarboxylate, 97%, Thermo Scientific Chemicals
Diethyl acetylenedicarboxylate, 97%, Thermo Scientific Chemicals
Diethyl acetylenedicarboxylate, 97%, Thermo Scientific Chemicals
Diethyl acetylenedicarboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethyl acetylenedicarboxylate, 97%, Thermo Scientific Chemicals
CAS: 762-21-0 | C8H10O4 | 170.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA296351000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black
Platinum (Pt)9.8 to 10.2 %
Appearance (Form)Powder
Moisture45 % to 60 %