Cyclopentene, 95+%, Thermo Scientific Chemicals
Cyclopentene, 95+%, Thermo Scientific Chemicals
Cyclopentene, 95+%, Thermo Scientific Chemicals
Cyclopentene, 95+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclopentene, 95+%, Thermo Scientific Chemicals
CAS: 142-29-0 | C5H8 | 68.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Packaging:
Glass bottle
Catalog number FSA295905000
Price (MYR)
1,302.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,302.00
Add to cart
Specifications
Chemical Name or MaterialCyclopentene
Name Notetech., ≥95%
CAS142-29-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Highly flammable liquid and vapor.