Phosphoric acid, extra pure, 85% solution in water, Thermo Scientific Chemicals
Phosphoric acid, extra pure, 85% solution in water, Thermo Scientific Chemicals
Phosphoric acid, extra pure, 85% solution in water, Thermo Scientific Chemicals
Phosphoric acid, extra pure, 85% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phosphoric acid, extra pure, 85% solution in water, Thermo Scientific Chemicals
CAS: 7664-38-2 | H3O4P | 97.994 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number FSA295700010
Price (MYR)
518.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
518.00
Add to cart
Chemical Identifiers
CAS555-57-7
IUPAC Namebenzyl(methyl)(prop-2-yn-1-yl)amine
Molecular FormulaC11H13N
InChI KeyDPWPWRLQFGFJFI-UHFFFAOYSA-N
SMILESCN(CC#C)CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO4>=96.0 %
Refractive index1.5210 to 1.5230 (20°C, 589 nm)
Appearance (Form)Liquid
Appearance (Color)Clear yellow