Methyl sulfoxide, 99.8+%, extra pure, Thermo Scientific Chemicals
Methyl sulfoxide, 99.8+%, extra pure, Thermo Scientific Chemicals
Methyl sulfoxide, 99.8+%, extra pure, Thermo Scientific Chemicals
Methyl sulfoxide, 99.8+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl sulfoxide, 99.8+%, extra pure, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA295520025
Price (MYR)
1,283.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,283.00
Add to cart
Specifications
Chemical Name or MaterialMethyl sulfoxide
Melting Point18.4°C
CAS67-68-5
Acidity0.04mg KOH/g max.
Density1.1000g/mL