Sebacoyl chloride, 92%, technical, Thermo Scientific Chemicals
Sebacoyl chloride, 92%, technical, Thermo Scientific Chemicals
Sebacoyl chloride, 92%, technical, Thermo Scientific Chemicals
Sebacoyl chloride, 92%, technical, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sebacoyl chloride, 92%, technical, Thermo Scientific Chemicals
Sebacoyl chloride, 92%, C10H16Cl2O2, CAS Number-111-19-3, decanedioic acid chloride, sebacoyl chloride, cp, octane-1,8-dicarbonyl chloride, sebacyl chloride, sebacoyl dichloride, sebacic acid dichloride, sebacoyl chloride, sebacoylchloride, acmc-1byrk, decanedioyl chloride, 500g, 90% min. (GC)
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
50 g
250 g
Packaging:
Glass bottle
Catalog number FSA294905000
Price (MYR)
2,439.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,439.00
Add to cart
Chemical Identifiers
CAS10025-77-1
IUPAC Nameiron(3+) hexahydrate trichloride
Molecular FormulaCl3FeH12O6
InChI KeyNQXWGWZJXJUMQB-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange to brown
Insoluble matter=<0.05 % (10 g in 50 ml H2O:HCl (49:1))
Sulfate (SO4)=<1500 ppm
Titration Iodometric99.0 to 102.0 %
Ferrous iron (Fe 2+)=<0.9 %