3-Cyclohexenecarboxylic acid, 97%, Thermo Scientific Chemicals
3-Cyclohexenecarboxylic acid, 97%, Thermo Scientific Chemicals
3-Cyclohexenecarboxylic acid, 97%, Thermo Scientific Chemicals
3-Cyclohexenecarboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Cyclohexenecarboxylic acid, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
Packaging:
Glass bottle
Catalog number FSA294390100
Price (MYR)
2,110.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,110.00
Add to cart
Specifications
Chemical Name or Material3-Cyclohexenecarboxylic acid
Name Note97%
CAS4771-80-6
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger