Cyclen, 97%, Thermo Scientific Chemicals
Cyclen, 97%, Thermo Scientific Chemicals
Cyclen, 97%, Thermo Scientific Chemicals
Cyclen, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclen, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
250 mg
1 g
Packaging:
Glass bottle
Catalog number FSA294320500
Price (MYR)
262.00
Add to cart
Quantity:
50 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
262.00
Add to cart
Specifications
Chemical Name or MaterialCyclen
Name Note97%
CAS294-90-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
View more
Chelation/Complexation reagent
RUO – Research Use Only