Cyclen, 97%, Thermo Scientific Chemicals
Cyclen, 97%, Thermo Scientific Chemicals
Cyclen, 97%, Thermo Scientific Chemicals
Cyclen, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclen, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mg
250 mg
Packaging:
Catalog number FSA294320010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Chemical Identifiers
CAS7782-68-5
IUPAC Namelithium(1+) iodate
Molecular FormulaILiO3
InChI KeyFZAXZVHFYFGNBX-UHFFFAOYSA-M
SMILES[Li+].[O-][I](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentIodide P.T.
Impurity contentNitrogen compounds (as N) 0.1%
Impurity contentSO₄ 0.015%
Assay (unspecified)99.5% HIO₃ min
Residue on ignition0.02%
View more
Chelation/Complexation reagent
RUO – Research Use Only