Trifluoroacetic acid, 99.5%, for biochemistry, Thermo Scientific Chemicals
Trifluoroacetic acid, 99.5%, for biochemistry, Thermo Scientific Chemicals
Trifluoroacetic acid, 99.5%, for biochemistry, Thermo Scientific Chemicals
Trifluoroacetic acid, 99.5%, for biochemistry, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trifluoroacetic acid, 99.5%, for biochemistry, Thermo Scientific Chemicals
CAS: 76-05-1 | C2HF3O2 | 114.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA293811000
Price (MYR)
527.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
527.00
Add to cart
Chemical Identifiers
CAS103-63-9
IUPAC Name(2-bromoethyl)benzene
Molecular FormulaC8H9Br
InChI KeyWMPPDTMATNBGJN-UHFFFAOYSA-N
SMILESBrCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5553 to 1.5573 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Water=<0.05 %
GC>=97.5 %
View more
For biochemistry
RUO – Research Use Only