Trifluoroacetic acid, 99.5%, for biochemistry, Thermo Scientific Chemicals
Trifluoroacetic acid, 99.5%, for biochemistry, Thermo Scientific Chemicals
Trifluoroacetic acid, 99.5%, for biochemistry, Thermo Scientific Chemicals
Trifluoroacetic acid, 99.5%, for biochemistry, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Trifluoroacetic acid, 99.5%, for biochemistry, Thermo Scientific Chemicals
CAS: 76-05-1 | C2HF3O2 | 114.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA293810250
Price (MYR)
298.00
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
298.00
Add to cart
Specifications
Chemical Name or MaterialTrifluoroacetic acid
CAS76-05-1
Melting Point-15.0°C
Boiling Point72.0°C
Health Hazard 1Danger
View more
For biochemistry
RUO – Research Use Only