Bis(diphenylphosphino)methane, 98%, Thermo Scientific Chemicals
Bis(diphenylphosphino)methane, 98%, Thermo Scientific Chemicals
Bis(diphenylphosphino)methane, 98%, Thermo Scientific Chemicals
Bis(diphenylphosphino)methane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Bis(diphenylphosphino)methane, 98%, Thermo Scientific Chemicals
CAS: 2071-20-7 | C25H22P2 | 384.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA293610050
Price (MYR)
600.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
600.00
Add to cart
Chemical Identifiers
CAS18282-59-2
IUPAC Name4-bromo-1,2-dichlorobenzene
Molecular FormulaC6H3BrCl2
InChI KeyCFPZDVAZISWERM-UHFFFAOYSA-N
SMILESClC1=CC=C(Br)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormClear liquid or fused solid
Refractive Index1.5975-1.6015 @ 20°C
Appearance (Color)Colorless or white to pale yellow
Assay (GC)≥98.0%
FormClear liquid or fused solid