4-Cyanostyrene, 95%, stabilized, Thermo Scientific Chemicals
4-Cyanostyrene, 95%, stabilized, Thermo Scientific Chemicals
4-Cyanostyrene, 95%, stabilized, Thermo Scientific Chemicals
4-Cyanostyrene, 95%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Cyanostyrene, 95%, stabilized, Thermo Scientific Chemicals
CAS: 3435-51-6 | C9H7N | 129.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
Packaging:
Glass bottle
Catalog number FSA292910025
Price (MYR)
1,115.00
Add to cart
Quantity:
2.5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,115.00
Add to cart
Chemical Identifiers
CAS75-57-0
IUPAC Nametetramethylazanium chloride
Molecular FormulaC4H12ClN
InChI KeyOKIZCWYLBDKLSU-UHFFFAOYSA-M
SMILES[Cl-].C[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥96.0%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)≤1.0%