Octyl 4-methoxycinnamate, 98%, stabilized, Thermo Scientific Chemicals
Octyl 4-methoxycinnamate, 98%, stabilized, Thermo Scientific Chemicals
Octyl 4-methoxycinnamate, 98%, stabilized, Thermo Scientific Chemicals
Octyl 4-methoxycinnamate, 98%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Octyl 4-methoxycinnamate, 98%, stabilized, Thermo Scientific Chemicals
CAS: 5466-77-3 | C18H26O3 | 290.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA291161000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS50926-11-9
IUPAC Namestannanone indium
Molecular FormulaInOSn
InChI KeyAMGQUBHHOARCQH-UHFFFAOYSA-N
SMILES[In].O=[Sn]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.01% max.