2-Bromoacetamide, 98%, Thermo Scientific Chemicals
2-Bromoacetamide, 98%, Thermo Scientific Chemicals
2-Bromoacetamide, 98%, Thermo Scientific Chemicals
2-Bromoacetamide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Bromoacetamide, 98%, Thermo Scientific Chemicals
CAS: 683-57-8 | C2H4BrNO | 137.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA291100050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS10344-94-2
IUPAC Name(2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-nitrophenoxy)oxane-2-carboxylate
Molecular FormulaC12H12NO9
InChI KeyQSUILVWOWLUOEU-GOVZDWNOSA-M
SMILESO[C@@H]1[C@@H](O)[C@H](OC2=CC=C(C=C2)[N+]([O-])=O)O[C@@H]([C@H]1O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Water=<8 % (K.F.)
Infrared spectrumConforms
HPLC=<0.05 % (p-Nitrophenol)
Solubility(1 % in water) Clear colorless to light yellow