Calix[4]arene, 98%, Thermo Scientific Chemicals
Calix[4]arene, 98%, Thermo Scientific Chemicals
Calix[4]arene, 98%, Thermo Scientific Chemicals
Calix[4]arene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Calix[4]arene, 98%, Thermo Scientific Chemicals
CAS: 74568-07-3 | C28H24O4 | 424.5 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA290190010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5351-23-5
IUPAC Name4-hydroxybenzohydrazide
Molecular FormulaC7H8N2O2
InChI KeyZMZGIVVRBMFZSG-UHFFFAOYSA-N
SMILESNNC(=O)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to pale grey
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder
Assay (HPLC)≥97.5%
Chelation/Complexation reagent
RUO – Research Use Only