Polysorbate 80, Thermo Scientific Chemicals
Polysorbate 80, Thermo Scientific Chemicals
Polysorbate 80, Thermo Scientific Chemicals
Polysorbate 80, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Polysorbate 80, Thermo Scientific Chemicals
Polysorbate 80, CAS Number-9005-65-6, Polyoxyethylene(20)sorbitan monooleate, 2.5L, 2mg KOH/g max., Yellow to Amber, 1.0800g/mL, 5ppm max., >110 deg.C, 65 to 80mg KOH/g, Authentic, 15, 7703, MFCD00082107, Glass bottle, Viscous Liquid At 25 deg.C, Indoors +20 deg.C, 1.4710 to 1.4730, 45 to 55mg KOH/g
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
250 mL
1 L
2.5 L
Catalog number FSA278630100
Price (MYR)
3,653.00
Add to cart
Quantity:
10 L
Request bulk or custom format
Price (MYR)
3,653.00
Add to cart
Chemical Identifiers
CAS10160-87-9
IUPAC Name3,3-diethoxyprop-1-yne
Molecular FormulaC7H12O2
InChI KeyRGUXEWWHSQGVRZ-UHFFFAOYSA-N
SMILESCCOC(OCC)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Refractive index1.4110 to 1.4140 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
RUO – Research Use Only
Contents & Storage
Indoors +20°C