Polysorbate 80, Thermo Scientific Chemicals
Polysorbate 80, Thermo Scientific Chemicals
Polysorbate 80, Thermo Scientific Chemicals
Polysorbate 80, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Polysorbate 80, Thermo Scientific Chemicals
Polysorbate 80, CAS Number-9005-65-6, Polyoxyethylene(20)sorbitan monooleate, 2.5L, 2mg KOH/g max., Yellow to Amber, 1.0800g/mL, 5ppm max., >110 deg.C, 65 to 80mg KOH/g, Authentic, 15, 7703, MFCD00082107, Glass bottle, Viscous Liquid At 25 deg.C, Indoors +20 deg.C, 1.4710 to 1.4730, 45 to 55mg KOH/g
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
2.5 L
10 L
Catalog number FSA278630010
Price (MYR)
628.00
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
628.00
Add to cart
Chemical Identifiers
CAS609-71-2
IUPAC Name2-oxo-1,2-dihydropyridine-3-carboxylic acid
Molecular FormulaC6H5NO3
InChI KeyUEYQJQVBUVAELZ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CNC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
Titration with NaOH>=97.5 %
Melting point256°C to 262°C
RUO – Research Use Only
Contents & Storage
Indoors +20°C