Carbopol™ 940
Carbopol™ 940
Carbopol™ 940
Thermo Scientific Chemicals
Carbopol™ 940
CAS: 9003-01-04 | (C3H4O2)n | 72.06 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA278382500250 gPlastic bottle
Catalog number FSA278382500
Price (MYR)
1,656.00
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,656.00
Add to cart
Specifications
Chemical Name or MaterialCarbopol* 94
CAS9003-01-04
ColorWhite
Loss on Drying2% max.
Infrared SpectrumAuthentic