Petroleum ether, for analysis, boiling range 80-110°C, Thermo Scientific Chemicals
Petroleum ether, for analysis, boiling range 80-110°C, Thermo Scientific Chemicals
Petroleum ether, for analysis, boiling range 80-110°C, Thermo Scientific Chemicals
Petroleum ether, for analysis, boiling range 80-110°C, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Petroleum ether, for analysis, boiling range 80-110°C, Thermo Scientific Chemicals
CAS: 64742-49-0 | C6H14 | 86.178 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number FSA278230010
Price (MYR)
334.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
334.00
Add to cart
Chemical Identifiers
CAS99-61-6
IUPAC Name3-nitrobenzaldehyde
Molecular FormulaC7H5NO3
InChI KeyZETIVVHRRQLWFW-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=CC(C=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to pale yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥98.5%
Identification (FTIR)Conforms
Free acid (titration)≤0.75%
View more
For analysis
RUO – Research Use Only