2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
CAS: 10312-55-7 | C8H5NO4 | 179.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA278030250
Price (MYR)
667.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
667.00
Add to cart
Chemical Identifiers
CAS395-01-7
IUPAC Name2,2-difluoro-1-phenylethan-1-one
Molecular FormulaC8H6F2O
InChI KeyOLYKCPDTXVZOQF-UHFFFAOYSA-N
SMILESFC(F)C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow or pale pink
Assay (GC)≥94.0%
FormLiquid
Refractive Index1.4950-1.5010 @ 20°C