1,6-Anhydro-β-D-glucopyranose, 99+%
1,6-Anhydro-β-D-glucopyranose, 99+%
1,6-Anhydro-β-D-glucopyranose, 99+%
1,6-Anhydro-β-D-glucopyranose, 99+%
Thermo Scientific Chemicals
1,6-Anhydro-β-D-glucopyranose, 99+%
CAS: 498-07-7 | C6H10O5 | 162.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA270950010
Price (MYR)
449.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
449.00
Add to cart
Chemical Identifiers
CAS149-57-5
IUPAC Name2-ethylhexanoic acid
Molecular FormulaC8H16O2
InChI KeyOBETXYAYXDNJHR-UHFFFAOYNA-N
SMILESCCCCC(CC)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<15 APHA
GC>=98.5 %
Water=<0.1 % (K.F.)