Alginic acid
Alginic acid
Alginic acid
Thermo Scientific Chemicals
Alginic acid
CAS: 9005-32-7 | (C6H8O7)A(C6H8O7)B
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number FSA270581000
Price (MYR)
456.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
456.00
Add to cart
Chemical Identifiers
CAS31786-45-5
IUPAC Name2-bromo-6-methoxybenzoic acid
Molecular FormulaC8H7BrO3
InChI KeyBNYFDICJYCBPRW-UHFFFAOYSA-N
SMILESCOC1=C(C(O)=O)C(Br)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)123.0-128.0?C
Appearance (Color)Pale cream to cream
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%