1,4-Dioxane, 99.5%, for HPLC, Thermo Scientific Chemicals
1,4-Dioxane, 99.5%, for HPLC, Thermo Scientific Chemicals
1,4-Dioxane, 99.5%, for HPLC, Thermo Scientific Chemicals
1,4-Dioxane, 99.5%, for HPLC, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,4-Dioxane, 99.5%, for HPLC, Thermo Scientific Chemicals
CAS: 123-91-1 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number FSA268340010
Price (MYR)
1,518.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,518.00
Add to cart
Chemical Identifiers
CAS1588-83-6
IUPAC Name4-amino-3-nitrobenzoic acid
Molecular FormulaC7H6N2O4
InChI KeyZZNAYFWAXZJITH-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1[N+]([O-])=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Yellow
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
Assay (HPLC)≥96.0%
For HPLC
RUO – Research Use Only