Acetonitrile, 99.9%, for HPLC, far UV, conform Eur.Ph., Thermo Scientific Chemicals
Acetonitrile, 99.9%, for HPLC, far UV, conform Eur.Ph., Thermo Scientific Chemicals
Acetonitrile, 99.9%, for HPLC, far UV, conform Eur.Ph., Thermo Scientific Chemicals
Acetonitrile, 99.9%, for HPLC, far UV, conform Eur.Ph., Thermo Scientific Chemicals
Thermo Scientific Chemicals
Acetonitrile, 99.9%, for HPLC, far UV, conform Eur.Ph., Thermo Scientific Chemicals
CAS: 75-05-8 | C2H3N | 41.053 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Glass Bottle
Catalog number FSA268260025
Price (MYR)
1,668.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,668.00
Add to cart
Chemical Identifiers
CAS9004-54-0
IUPAC NameDextran
Molecular Formula(C6H9O5)n(C6H10O5)m
SMILESO[C@@H]1[C@@H](O)[C@@H](-*)O[C@H](CO[C@H]2O[C@H](CO-*)[C@@H](O)[C@H](O-*)[C@H]2O)[C@H]1O
MDL NumberMFCD00130935
SpecificationsSpecification SheetSpecification Sheet
FormWhite to off white powder
Assay (unspecified)Specific rotation (2% in water) [|b]: NLT + 148?
Loss on Drying≤ 7.0 %
pH4.5 to 7.0 (10% solution)
Solution Test1% in water
View more
For HPLC
RUO – Research Use Only