Molecular sieves 4A, 10 to 18 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 10 to 18 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 10 to 18 mesh, Thermo Scientific Chemicals
Molecular sieves 4A, 10 to 18 mesh, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Molecular sieves 4A, 10 to 18 mesh, Thermo Scientific Chemicals
CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA264755000
Price (MYR)
579.00
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
579.00
Add to cart
Chemical Identifiers
CAS34202-69-2
IUPAC Namehexafluoropropan-2-one trihydrate
Molecular FormulaC3H6F6O4
InChI KeySNZAEUWCEHDROX-UHFFFAOYSA-N
SMILESO.O.O.FC(F)(F)C(=O)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.3160-1.3220 @ 20°C
FormLiquid
Water Content (Karl Fischer Titration)21.0-30.0% (2.4-3.4 waters of hydration)
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Drying agent
RUO – Research Use Only