Sodium hydroxide, extra pure, 50 wt% solution in water, Thermo Scientific Chemicals
Sodium hydroxide, extra pure, 50 wt% solution in water, Thermo Scientific Chemicals
Sodium hydroxide, extra pure, 50 wt% solution in water, Thermo Scientific Chemicals
Sodium hydroxide, extra pure, 50 wt% solution in water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium hydroxide, extra pure, 50 wt% solution in water, Thermo Scientific Chemicals
CAS: 1310-73-2 | HNaO | 39.997 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Plastic Bottle
Catalog number FSA259860010
Price (MYR)
381.00
Add to cart
Quantity:
1 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
381.00
Add to cart
Specifications
Chemical Name or MaterialSodium hydroxide
ColorWhite to Gray
Name Note50 wt% solution in water
Boiling Point145.0°C
CAS1310-73-2