Butyl butyrate, 98%, Thermo Scientific Chemicals
Butyl butyrate, 98%, Thermo Scientific Chemicals
Butyl butyrate, 98%, Thermo Scientific Chemicals
Butyl butyrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Butyl butyrate, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number FSA259591000
Price (MYR)
205.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
205.00
Add to cart
Chemical Identifiers
CAS78-82-0
IUPAC Name2-methylpropanenitrile
Molecular FormulaC4H7N
InChI KeyLRDFRRGEGBBSRN-UHFFFAOYSA-N
SMILESCC(C)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.3720 to 1.3740 (20°C, 589 nm)
Appearance (Form)Liquid