1,10-Dichlorodecane, 98%
1,10-Dichlorodecane, 98%
1,10-Dichlorodecane, 98%
1,10-Dichlorodecane, 98%
Thermo Scientific Chemicals
1,10-Dichlorodecane, 98%
CAS: 2162-98-3 | C10H20Cl2 | 211.17 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA25593025025 gGlass bottle
Catalog number FSA255930250
Price (MYR)
1,250.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,250.00
Add to cart
Chemical Identifiers
CAS58880-37-8
IUPAC Name1-(4-chlorophenyl)cyclohexane-1-carboxylate
Molecular FormulaC13H14ClO2
InChI KeyUPNXUJXIIZGXLQ-UHFFFAOYSA-M
SMILES[O-]C(=O)C1(CCCCC1)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥94.0 to ≤106.0%
Appearance (Color)White to grey or purple
FormPowder or granules
Identification (FTIR)Conforms
Melting Point (clear melt)146.5-158.5?C