3-Chloroperoxybenzoic acid, 70-75%, balance 3-Chlorobenzoic acid and water, Thermo Scientific Chemicals
3-Chloroperoxybenzoic acid, 70-75%, balance 3-Chlorobenzoic acid and water, Thermo Scientific Chemicals
3-Chloroperoxybenzoic acid, 70-75%, balance 3-Chlorobenzoic acid and water, Thermo Scientific Chemicals
3-Chloroperoxybenzoic acid, 70-75%, balance 3-Chlorobenzoic acid and water, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Chloroperoxybenzoic acid, 70-75%, balance 3-Chlorobenzoic acid and water, Thermo Scientific Chemicals
3-Chloroperoxybenzoic acid, 70-75%, C7H5ClO3, CAS Number-937-14-4, 535-80-8, 7732-18-5
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
View all
Plastic Bottle
Catalog number FSA255790250
Price (MYR)
420.00
Add to cart
Quantity:
25 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
420.00
Add to cart
Chemical Identifiers
CAS17481-27-5
IUPAC Name3-amino-4-methoxybenzamide
Molecular FormulaC8H10N2O2
InChI KeyINCJNDAQNPWMPZ-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C1N)C(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to pale brown or pale pink
Assay (GC)≥97.5%
Melting Point (clear melt)154.0-161.0?C
FormPowder
Identification (FTIR)Conforms
Oxidation reagent
RUO – Research Use Only