Sodium hyaluronate, 95%, Thermo Scientific Chemicals
Sodium hyaluronate, 95%, Thermo Scientific Chemicals
Sodium hyaluronate, 95%, Thermo Scientific Chemicals
Sodium hyaluronate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium hyaluronate, 95%, Thermo Scientific Chemicals
CAS: 9067-32-7 | (C14H20NO11Na)n | 417.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number FSA251770050
Price (MYR)
1,638.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,638.00
Add to cart
Chemical Identifiers
CAS22053-74-3
IUPAC Name3-methyl-1-benzothiophene-2-carbaldehyde
Molecular FormulaC10H8OS
InChI KeyDRZGHNXLEQHVHB-UHFFFAOYSA-N
SMILESCC1=C(SC2=CC=CC=C12)C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Free acid (titration)<1.0%
Assay (GC)>97.5%
Melting Point85-89? (with ≤3? range)