Sodium hyaluronate, 95%, Thermo Scientific Chemicals
Sodium hyaluronate, 95%, Thermo Scientific Chemicals
Sodium hyaluronate, 95%, Thermo Scientific Chemicals
Sodium hyaluronate, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Sodium hyaluronate, 95%, Thermo Scientific Chemicals
CAS: 9067-32-7 | (C14H20NO11Na)n | 417.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA251770010
Price (MYR)
502.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
502.00
Add to cart
Chemical Identifiers
CAS35486-42-1
IUPAC Name3,5-dibromopyridin-2-amine
Molecular FormulaC5H4Br2N2
InChI KeyWJMJWMSWJSACSN-UHFFFAOYSA-N
SMILESNC1=NC=C(Br)C=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow or cream or pale brown to brown
FormCrystals or Crystalline powder
Assay (GC)≥96.0%
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
Melting Point (clear melt)100-106?C