L-Lactic acid, sodium salt, 60 wt% solution in water
L-Lactic acid, sodium salt, 60 wt% solution in water
L-Lactic acid, sodium salt, 60 wt% solution in water
L-Lactic acid, sodium salt, 60 wt% solution in water
Thermo Scientific Chemicals
L-Lactic acid, sodium salt, 60 wt% solution in water
CAS: 867-56-1 | C3H5NaO3 | 112.06 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA2503000101 LGlass bottle
Catalog number FSA250300010
Price (MYR)
521.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
521.00
Add to cart
Chemical Identifiers
CAS867-56-1, 64-17-5
Specifications
Chemical Name or MaterialL-Lactic Acid, Sodium Salt
Name Note60 wt% Solution in Water
CAS867-56-1
Health Hazard 1
CAS64-17-5