Calcium L-lactate pentahydrate, 98%
Calcium L-lactate pentahydrate, 98%
Calcium L-lactate pentahydrate, 98%
Calcium L-lactate pentahydrate, 98%
Thermo Scientific Chemicals
Calcium L-lactate pentahydrate, 98%
CAS: 865717-20-0 | C6H10CaO6·5H2O | 308.29 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA2502000101 kgPlastic bottle
Catalog number FSA250200010
Price (MYR)
423.00
Add to cart
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
423.00
Add to cart
Chemical Identifiers
CAS75-59-2
IUPAC Nametetramethylazanium hydroxide
Molecular FormulaC4H13NO
InChI KeyWGTYBPLFGIVFAS-UHFFFAOYSA-M
SMILES[OH-].C[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Titration with HCl23 to 27 %
Refractive index1.3780 to 1.3840 (20°C, 589 nm)
Appearance (Form)Solution