n-Butyl lactate, 99%, Thermo Scientific Chemicals
n-Butyl lactate, 99%, Thermo Scientific Chemicals
n-Butyl lactate, 99%, Thermo Scientific Chemicals
n-Butyl lactate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Butyl lactate, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA250185000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1878-49-5
IUPAC Name2-(2-methylphenoxy)acetic acid
Molecular FormulaC9H10O3
InChI KeyQJVXBRUGKLCUMY-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1OCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale cream
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (GC)≥97.5%
Melting Point (clear melt)152.0-158.0?C