n-Butyl lactate, 99%, Thermo Scientific Chemicals
n-Butyl lactate, 99%, Thermo Scientific Chemicals
n-Butyl lactate, 99%, Thermo Scientific Chemicals
n-Butyl lactate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
n-Butyl lactate, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
500 mL
Packaging:
Glass bottle
Catalog number FSA250180025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS103-33-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to red
Assay (GC)≥97.0%
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder or chunks
Melting Point (clear melt)62.0-72.0?C