1-Chloro-2-iodobenzene, 99%, Thermo Scientific Chemicals
1-Chloro-2-iodobenzene, 99%, Thermo Scientific Chemicals
1-Chloro-2-iodobenzene, 99%, Thermo Scientific Chemicals
1-Chloro-2-iodobenzene, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1-Chloro-2-iodobenzene, 99%, Thermo Scientific Chemicals
CAS: 615-41-8 | C6H4ClI | 238.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number FSA249460500
Price (MYR)
1,225.00
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,225.00
Add to cart
Chemical Identifiers
CAS35963-20-3
IUPAC Name[(1R)-7,7-dimethyl-2-oxobicyclo[2.2.1]heptan-1-yl]methanesulfonic acid
Molecular FormulaC10H16O4S
InChI KeyMIOPJNTWMNEORI-MHPPCMCBSA-N
SMILESCC1(C)C2CC[C@]1(CS(O)(=O)=O)C(=O)C2
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration with NaOH>=97.5 %
Appearance (Color)White to light beige
Infrared spectrumConforms
Specific optical rotation-21° to -23° (20°C, 589 nm) (c=20, H2O)