3-Thiophenecarboxylic acid, 99%, Thermo Scientific Chemicals
3-Thiophenecarboxylic acid, 99%, Thermo Scientific Chemicals
3-Thiophenecarboxylic acid, 99%, Thermo Scientific Chemicals
3-Thiophenecarboxylic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3-Thiophenecarboxylic acid, 99%, Thermo Scientific Chemicals
CAS: 88-13-1 | C5H4O2S | 128.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA243110010
Price (MYR)
285.00
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
285.00
Add to cart
Chemical Identifiers
CAS108-40-7
IUPAC Name3-methylbenzene-1-thiol
Molecular FormulaC7H8S
InChI KeyWRXOZRLZDJAYDR-UHFFFAOYSA-N
SMILESCC1=CC=CC(S)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5735-1.5785 @ 20?C
Appearance (Color)Clear colorless to pale yellow