Phloroglucinol, 99+%, anhydrous, Thermo Scientific Chemicals
Phloroglucinol, 99+%, anhydrous, Thermo Scientific Chemicals
Phloroglucinol, 99+%, anhydrous, Thermo Scientific Chemicals
Phloroglucinol, 99+%, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Phloroglucinol, 99+%, anhydrous, Thermo Scientific Chemicals
CAS: 108-73-6 | C6H6O3 | 126.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number FSA241761000
Price (MYR)
660.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
660.00
Add to cart
Chemical Identifiers
CAS34369-07-8
IUPAC Namedisodium phosphono ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate
Molecular FormulaC10H14N5Na2O13P3
InChI KeyTTWYZDPBDWHJOR-WCYUCLFNNA-L
SMILES[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormPowder or crystals or crystalline powder
Assay (unspecified)≥97.5%
Water Content (Karl Fischer Titration)10% max.