Silica gel, for column chromatography, 0.200-0.500mm, 40Å
Silica gel, for column chromatography, 0.200-0.500mm, 40Å
Silica gel, for column chromatography, 0.200-0.500mm, 40Å
Silica gel, for column chromatography, 0.200-0.500mm, 40Å
Silica gel, for column chromatography, 0.200-0.500mm, 40Å
Thermo Scientific Chemicals
Silica gel, for column chromatography, 0.200-0.500mm, 40Å
CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number FSA241670010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Chemical Identifiers
CAS100-51-6
IUPAC Namephenylmethanol
Molecular FormulaC7H8O
InChI KeyWVDDGKGOMKODPV-UHFFFAOYSA-N
SMILESOCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance of solutionClear colorless
Peroxide value=<5
Residue after evaporation=<0.05 %
Related substances=<0.04 (Sum of other peaks with relative retention
View more
For column chromatography
RUO – Research Use Only