Silica gel, for chromatography, 0.200-0.500mm, 60Å
Silica gel, for chromatography, 0.200-0.500mm, 60Å
Silica gel, for chromatography, 0.200-0.500mm, 60Å
Silica gel, for chromatography, 0.200-0.500mm, 60Å
Thermo Scientific Chemicals
Silica gel, for chromatography, 0.200-0.500mm, 60Å
Have Questions?
Change viewbuttonViewtableView
Packaging:
Plastic Bottle
Quantity:
1 kg
Catalog number FSA240380010
Price (MYR)
612.00
Add to cart
Packaging:
Plastic Bottle
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
612.00
Add to cart
Chemical Identifiers
CAS294-90-6
IUPAC Name1,4,7,10-tetraazacyclododecane
Molecular FormulaC8H20N4
InChI KeyQBPPRVHXOZRESW-UHFFFAOYSA-N
SMILESC1CNCCNCCNCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Melting point108°C to 113°C
GC>=96.0 %
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Applications:
For column chromatography
RUO – Research Use Only