Silica Gel, for Column Chromatography, 0.060-0.200mm, 60Å
Silica Gel, for Column Chromatography, 0.060-0.200mm, 60Å
Silica Gel, for Column Chromatography, 0.060-0.200mm, 60Å
Silica Gel, for Column Chromatography, 0.060-0.200mm, 60Å
Silica Gel, for Column Chromatography, 0.060-0.200mm, 60Å
Thermo Scientific Chemicals
Silica Gel, for Column Chromatography, 0.060-0.200mm, 60Å
For column chromatography.
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
30 kg
Catalog number FSA240370050
Price (MYR)
2,792.00
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (MYR)
2,792.00
Add to cart
Chemical Identifiers
CAS32955-21-8
IUPAC Nameethyl 2-amino-1,3-thiazole-5-carboxylate
Molecular FormulaC6H8N2O2S
InChI KeyVNZXERIGKZNEKB-UHFFFAOYSA-N
SMILESCCOC(=O)C1=CN=C(N)S1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)Yellow
Melting Point (clear melt)157.0-166.0?C
Assay (GC)≥96.0%