Silica gel, for column chromatography, 0.035-0.070mm, 60Å
Silica gel, for column chromatography, 0.035-0.070mm, 60Å
Silica gel, for column chromatography, 0.035-0.070mm, 60Å
Silica gel, for column chromatography, 0.035-0.070mm, 60Å
Silica gel, for column chromatography, 0.035-0.070mm, 60Å
Thermo Scientific Chemicals
Silica gel, for column chromatography, 0.035-0.070mm, 60Å
For column chromatography.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Catalog number FSA240360010
Price (MYR)
775.00
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
775.00
Add to cart
Chemical Identifiers
CAS1684-40-8
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Purity≥97.0%
FormPowder or crystals or crystalline powder
For column chromatography
RUO – Research Use Only