Butylated hydroxyanisole, 96%, Thermo Scientific Chemicals
Butylated hydroxyanisole, 96%, Thermo Scientific Chemicals
Butylated hydroxyanisole, 96%, Thermo Scientific Chemicals
Butylated hydroxyanisole, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Butylated hydroxyanisole, 96%, Thermo Scientific Chemicals
CAS: 25013-16-5 | C11H16O2 | 180.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number FSA235231000
Price (MYR)
318.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
318.00
Add to cart
Chemical Identifiers
CAS5576-97-6
IUPAC Name1-methoxy-4-(2-nitroethenyl)benzene
Molecular FormulaC9H9NO3
InChI KeyJKQUXSHVQGBODD-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=C[N+]([O-])=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)84.5-90.5?C