Polysorbate 20, Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
CAS: 9005-64-5 | C26H50O10
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
5 mL
250 mL
1 L
5 L
Packaging Type:
View all
Plastic drum
Catalog number FSA233360250
Price (MYR)
4,061.00
Add to cart
Quantity:
25 L
Packaging Type:
Plastic drum
Request bulk or custom format
Price (MYR)
4,061.00
Add to cart
Chemical Identifiers
CAS555-21-5
IUPAC Name2-(4-nitrophenyl)acetonitrile
Molecular FormulaC8H6N2O2
InChI KeyPXNJGLAVKOXITN-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(CC#N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5%
Melting Point (clear melt)111.0-118.0?C