Polysorbate 20, Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Polysorbate 20, Thermo Scientific Chemicals
CAS: 9005-64-5 | C26H50O10
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
5 mL
250 mL
1 L
25 L
Packaging Type:
View all
Plastic Bottle
Catalog number FSA233360051
Price (MYR)
1,344.00
Add to cart
Quantity:
5 L
Packaging Type:
Plastic Bottle
Request bulk or custom format
Price (MYR)
1,344.00
Add to cart
Chemical Identifiers
CAS51594-55-9
IUPAC Name(2R)-2-(chloromethyl)oxirane
Molecular FormulaC3H5ClO
InChI KeyBRLQWZUYTZBJKN-VKHMYHEASA-N
SMILESClC[C@H]1CO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
GC>=98.5 %
Infrared spectrumConforms
Specific optical rotation-34° to -39° (20°C, 589 nm) (c=1, MeOH)
Appearance (Form)Liquid